Learn More
Sino Biological Recombinant Human TMPRSS11B Protein (His Tag)
Supplier:  Sino Biological 15888H07H100
Encompass_Preferred
A DNA sequence encoding the human TMPRSS11B (CAD91168.1) (Leu39-Leu416) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9758