Learn More
CEM Corporation 159875 TRACK FILM 600SH/PK
SDP
Catalog No. NC9146845
Encompass
Trac Film (1 roll, 600 sheets). Trac film consists of a proprietary chemical composition which ensures an absolute minimum in interference for fat determination by NMR. CEM Corporation individually tests each batch to ensure that this standard is continuously met. Therefore, when using CEM’s Trac film you will ensure the best performance possible with your Trac system. Trac film, developed by CEM Corporation is protected by worldwide patents in the US, Canada, Europe, and Japan*. Use of any non-CEM film is in violation of these patents and will compromise the performance of the Trac system. This includes the performance of all methods transferred onto the system that were developed with CEM Trac film. Additionally, the use of a non-CEM film substitute may lead to voidance of the system warranty or service contract.
Catalog No. NC9146845 Supplier CEM Corporation Supplier No. 159875
Trac Film (1 roll, 600 sheets). Trac film consists of a proprietary chemical composition which ensures an absolute minimum in interference for fat determination by NMR. CEM Corporation individually tests each batch to ensure that this standard is continuously met. Therefore, when using CEM’s Trac film you will ensure the best performance possible with your Trac system. Trac film, developed by CEM Corporation is protected by worldwide patents in the US, Canada, Europe, and Japan*. Use of any non-CEM film is in violation of these patents and will compromise the performance of the Trac system. This includes the performance of all methods transferred onto the system that were developed with CEM Trac film. Additionally, the use of a non-CEM film substitute may lead to voidance of the system warranty or service contract.