Learn More
Sino Biological Recombinant Human IMPA1 Protein (His Tag)
Supplier:  Sino Biological 15998H07E100
Encompass_Preferred
A DNA sequence encoding the human IMPA1 (P29218-1) (Met1-Asp277) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9772