Learn More
MSC Ferraz Shawmut TRS15R 300 VDC, 600 VAC, 15 Amp, Time Delay General Purpose Fuse Clip Mount, 127mm Overall Length, 20 at DC, 200 at AC kA Rating, 13/16" Diam
Catalog No. 501296476
Encompass
Ferraz Shawmut TRS15R 300 VDC, 600 VAC, 15 Amp, Time Delay General Purpose Fuse Clip Mount, 127mm Overall Length, 20 at DC, 200 at AC kA Rating, 13/16" Diam
Catalog No. 50-129-6476 Supplier MSC Supplier No. 54006424
Ferraz Shawmut TRS15R 300 VDC, 600 VAC, 15 Amp, Time Delay General Purpose Fuse Clip Mount, 127mm Overall Length, 20 at DC, 200 at AC kA Rating, 13/16" Diam