Learn More
MSC Cooper Bussmann FRS-R-15ID 300 VDC, 600 VAC, 15 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 20.6mm Diam
Supplier:  MSC 91700336
Encompass
Cooper Bussmann FRS-R-15ID 300 VDC, 600 VAC, 15 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 20.6mm Diam
Catalog No. 50-129-6474