Learn More
Grainger MERSEN, Fuse, 15A 5PK
Catalog No. NC2074475
Encompass_Preferred
The Mersen 15 Amp Fuse is a fast-acting, current-limiting fuse designed to protect electrical circuits and equipment. This UL Listed Class CC fuse has a voltage rating of 600VAC and features nickel-plated brass end caps with a glass tube body. The compact 1-1/4" x 1/4" cylindrical fuse body allows for use in tight spaces. With a 15A amperage rating, this fuse provides overload and short circuit protection for a variety of applications including industrial controls, HVAC equipment, power distribution panels, and more. The fast-acting fuse limits damage by quickly opening the circuit during overcurrent conditions.
Catalog No. NC2074475 Supplier Grainger Supplier No. 5LCA2
The Mersen 15 Amp Fuse is a fast-acting, current-limiting fuse designed to protect electrical circuits and equipment. This UL Listed Class CC fuse has a voltage rating of 600VAC and features nickel-plated brass end caps with a glass tube body. The compact 1-1/4" x 1/4" cylindrical fuse body allows for use in tight spaces. With a 15A amperage rating, this fuse provides overload and short circuit protection for a variety of applications including industrial controls, HVAC equipment, power distribution panels, and more. The fast-acting fuse limits damage by quickly opening the circuit during overcurrent conditions.