Learn More
Med Vet International Sterile IV Administration Set, 15dr/mL, 100in with Injection Site, Luer Slip
Supplier:  Med Vet International IV15
Encompass_Preferred
Our IV administration sets feature universal kink-resistant tubing with smooth action roller clamp, and Y injection site. Available in 10, 15, 20 and 60 drop per mL Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Luer Slip Meets a wide range of solution administration requirements. Tubing Length: 100' Luer Slip Connection
Catalog No. NC0077014