Learn More
MSC Honeywell North Work Gloves, 15 Gauge, Nylon, PVC-Coated, Large
Catalog No. 5000509758
Encompass
This ultra-lightweight, general-purpose glove features a 15 gauge seamless nylon shell with a foamed PVC knuckle length coating. The foamed PVC provides an exceptional combination of abrasion resistance and grip, particularly in wet conditions, while maintaining dexterity and tactility. Its stretchy, form-fitting shell offers all-day comfort, and the PVC coating helps prevent liquid penetration and protects from knuckle abrasions.
  • Ultra-lightweight design for general purpose applications.
  • Foamed PVC coating offers exceptional abrasion resistance and grip, even in wet conditions.
  • Lightweight design maintains high dexterity and tactility.
  • Stretchy and form-fitting shell provides all-day comfort.
  • Bright red color enhances visibility.
  • Seamless nylon shell.
  • Knuckle-length PVC coating protects against liquid penetration and abrasions.
Catalog No. 50-005-09758 Supplier MSC Supplier No. 70523584
This ultra-lightweight, general-purpose glove features a 15 gauge seamless nylon shell with a foamed PVC knuckle length coating. The foamed PVC provides an exceptional combination of abrasion resistance and grip, particularly in wet conditions, while maintaining dexterity and tactility. Its stretchy, form-fitting shell offers all-day comfort, and the PVC coating helps prevent liquid penetration and protects from knuckle abrasions.
  • Ultra-lightweight design for general purpose applications.
  • Foamed PVC coating offers exceptional abrasion resistance and grip, even in wet conditions.
  • Lightweight design maintains high dexterity and tactility.
  • Stretchy and form-fitting shell provides all-day comfort.
  • Bright red color enhances visibility.
  • Seamless nylon shell.
  • Knuckle-length PVC coating protects against liquid penetration and abrasions.