Learn More
Cayman Chemical 15hydroxy PentadecNIc ACD 10mg
SDP
Catalog No. 502935750
Encompass_Preferred
A hydroxy fatty acid methyl ester; a key intermediate in the synthesis of the musk-odored macrocyclic lactones cyclopentadecanolide and exaltolide
Catalog No. 50-293-5750 Supplier Cayman Chemical Supplier No. 2464910 MG
A hydroxy fatty acid methyl ester; a key intermediate in the synthesis of the musk-odored macrocyclic lactones cyclopentadecanolide and exaltolide