Learn More
Cayman Chemical 15-hydroxy Pentadecanoic Acid, Purity: ≥98%, CAS: 4617-33-8, 10 mg
SDP
Catalog No. 502935729
Encompass_Preferred
A hydroxylated fatty acid; has been used in the synthesis of the macrocyclic lactone musk compounds exaltolide and phoracantholide I
Catalog No. 50-293-5729 Supplier Cayman Chemical Supplier No. 2464810 MG
A hydroxylated fatty acid; has been used in the synthesis of the macrocyclic lactone musk compounds exaltolide and phoracantholide I