Learn More
Andwin Scientific 15IN X13IN GRANITE BLOCK
SDP
Catalog No. NC9438955
Encompass_Preferred
The Granite platform measures 13" x 15". This comes with isolators, which are highgrade silicone connected to the platform and the base. The isolators filter out the vibrations to enhance the balance performance. This product is compatible with all MSA models, all Cubis Ultra-Micro, Micro Models, CPA26P and CPA225D.
Catalog No. NC9438955 Supplier Andwin Scientific Supplier No. 21201315
The Granite platform measures 13" x 15". This comes with isolators, which are highgrade silicone connected to the platform and the base. The isolators filter out the vibrations to enhance the balance performance. This product is compatible with all MSA models, all Cubis Ultra-Micro, Micro Models, CPA26P and CPA225D.