Learn More
Waters Corp WATERS CORPORATION Pump Head, 15K PSI, Stainless Steel
Catalog No. 50792772 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement pump head is constructed from stainless steel. It is designed for use on binary, isocratic, and quaternary solvent managers, as well as nano solvent managers.
  • Replacement stainless steel pump head.
  • Used on binary, isocratic, and quaternary solvent managers.
  • Used on nano solvent managers.
  • System type: UPLC.
  • Replacement schedule: As required.
Catalog No. 50-792-772 Supplier Waters Corp Supplier No. 700002595
This replacement pump head is constructed from stainless steel. It is designed for use on binary, isocratic, and quaternary solvent managers, as well as nano solvent managers.
  • Replacement stainless steel pump head.
  • Used on binary, isocratic, and quaternary solvent managers.
  • Used on nano solvent managers.
  • System type: UPLC.
  • Replacement schedule: As required.