Learn More
Cayman Chemical 15-keto Fluprostenol isopropyl ester, Purity: ≥98%, CAS: 404830-45-1, 10 mg
SDP
Catalog No. 502967749
Encompass_Preferred
An ester derivative of fluprostenol; a potential inactive metabolite of fluprostenol isopropyl ester (travoprost); a potential impurity found in commercial preparations of fluprostenol isopropyl ester
Catalog No. 50-296-7749 Supplier Cayman Chemical Supplier No. 1678610 MG
An ester derivative of fluprostenol; a potential inactive metabolite of fluprostenol isopropyl ester (travoprost); a potential impurity found in commercial preparations of fluprostenol isopropyl ester