Learn More
Ace Glass, Inc. Mantle, 10L/20L, for 230mm diameter cylindrical flask, 230mm ID x 69.9mm height, 250w-115v, bottom opening, CSA rated.
SDP
Catalog No. 50977336
Encompass
15L/20L aluminum housed, table-model mantle with bottom opening for 230mm diameter cylindrical flask. 230mm I.D. x 69.9mm height. Maximum internal operating temperature is 450?C. 250w-115v, and is cULus rated. Power cord included.
Catalog No. 50-977-336 Supplier Ace Glass, Inc. Supplier No. 1205364
15L/20L aluminum housed, table-model mantle with bottom opening for 230mm diameter cylindrical flask. 230mm I.D. x 69.9mm height. Maximum internal operating temperature is 450?C. 250w-115v, and is cULus rated. Power cord included.