Learn More
Ace Glass, Inc. Mantle, 15L, for 251mm diameter cylindrical flask, 251.5mm ID x 127mm height, 250w-115v, CSA rated
SDP
Catalog No. 50977332
Encompass
15L aluminum housed, table-model mantle for 251mm diameter cylindrical flask. 251.5mm I.D. x 127mm height. Maximum internal operating temperature is 450 Deg. centigrade. 250w-115v, cULus rated. Power cord included.
Catalog No. 50-977-332 Supplier Ace Glass, Inc. Supplier No. 1205343
15L aluminum housed, table-model mantle for 251mm diameter cylindrical flask. 251.5mm I.D. x 127mm height. Maximum internal operating temperature is 450 Deg. centigrade. 250w-115v, cULus rated. Power cord included.