Learn More
Chemglass Life Sciences 15mL Amber Centrifuge Tube, PS Rack, Sterile
SDP
Catalog No. 501897803
Encompass_Preferred
• Meet the requirements of analytical level • High strength non-toxic materials • Raw material complies with USP VI requirements • Printed clear graduations with marking area • PP for the tube and HDPE for the cap • One-hand operation design • Sterilized by E-Beam • DNase, RNase and pyrogen free • Maximum centrifugation speed 12,000 x g
Catalog No. 50-189-7803 Supplier Chemglass Life Sciences Supplier No. CGN4303A15
• Meet the requirements of analytical level • High strength non-toxic materials • Raw material complies with USP VI requirements • Printed clear graduations with marking area • PP for the tube and HDPE for the cap • One-hand operation design • Sterilized by E-Beam • DNase, RNase and pyrogen free • Maximum centrifugation speed 12,000 x g