Learn More
Bellco Glass, Inc. 15L Heating Blanket, 115V Bioreactor
SDP
Catalog No. 501946500
Encompass_Preferred
The blanket has velcro closures for easy setup and a 6" power cable with IEC 60320 C14 plug. 15L Heating Blanket 115V Bioreactor. MADE IN THE USA
Catalog No. 50-194-6500 Supplier Bellco Glass, Inc. Supplier No. 790925115
The blanket has velcro closures for easy setup and a 6" power cable with IEC 60320 C14 plug. 15L Heating Blanket 115V Bioreactor. MADE IN THE USA