Learn More
Ace Glass, Inc. Instatherm; oil bath, high form with rolled lip for easy handling, 15 liter, 290mm ID, 300mm OD, 240mm height, includes9698-20 connecting controller cord, 120v/14amp max
SDP
Catalog No. 5097758
Encompass
Catalog No. 50-977-58 Supplier Ace Glass, Inc. Supplier No. 960123