Learn More
Cayman Chemical 15-Lipoxygenase-2 Blocking Peptide, 1 ea
SDP
Catalog No. 502887055
Encompass_Preferred
Immunogen: Synthetic peptide from the internal region of 15-LO-2 · To be used in conjunction with Cayman’s 15-LO-2 polyclonal antibody (Catalog No. 10004454) to block protein-antibody complex formation during immunochemical analysis of 15-LO-2.
Catalog No. 50-288-7055 Supplier Cayman Chemical Supplier No. 100044571 EA
Immunogen: Synthetic peptide from the internal region of 15-LO-2 · To be used in conjunction with Cayman’s 15-LO-2 polyclonal antibody (Catalog No. 10004454) to block protein-antibody complex formation during immunochemical analysis of 15-LO-2.