Learn More
Chemglass Life Sciences 15mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 14/20 Outer Angled 20°
SDP
Catalog No. CG155806
Encompass_Preferred
15mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 14/20 Outer Angled 20¿. Pear shape, HEAVY WALL 2-neck flask with standard taper outer joints.
Catalog No. CG155806 Supplier Chemglass Life Sciences Supplier No. CG155806
15mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 14/20 Outer Angled 20¿. Pear shape, HEAVY WALL 2-neck flask with standard taper outer joints.