Learn More
Thomas Scientific Bottle, 1/2 oz/15 ml, 1000 Class, 24/case
SDP
Catalog No. NC0284299
Encompass
10 Class jars are supplied with Teflon /polypropylene caps and are ready for immediate use Offered pre-cleaned with or without Independent Laboratory analysis 3000 Class recommended for organic and inorganic analysis 1755G16 and G18 can be used with Lars Lande, Millipore and other box-type rotary TCLP agitators.
Catalog No. NC0284299 Supplier Thomas Scientific Supplier No. 1755G67
10 Class jars are supplied with Teflon /polypropylene caps and are ready for immediate use Offered pre-cleaned with or without Independent Laboratory analysis 3000 Class recommended for organic and inorganic analysis 1755G16 and G18 can be used with Lars Lande, Millipore and other box-type rotary TCLP agitators.