Learn More
Quality Biological Inc 15 mL Bio-Reaction Tube - Sterile, Bulk Pack, 10/Resealabe Bag, 100/CS
SDP
Catalog No. 50147452
Encompass_Preferred
15 mL Bio-Reaction Tube - Sterile (gamma irradiated). , Bulk Pack, 10/Resealabe Bag, 100/CS
Vented, hydrophobic membrane cap. For small scale suspension culture with gas exchange.
Catalog No. 50-147-452 Supplier Quality Biological Inc Supplier No. P886229471
15 mL Bio-Reaction Tube - Sterile (gamma irradiated). , Bulk Pack, 10/Resealabe Bag, 100/CS
Vented, hydrophobic membrane cap. For small scale suspension culture with gas exchange.