Learn More
Quality Biological Inc 15 mL Bio-Reaction Tube with Vented Cap - Sterile, Foam Rack, 50/Rack, 300/CS
SDP
Catalog No. 50147453
Encompass_Preferred
15 mL Bio-Reaction Tube with Vented Cap - Sterile, Foam Rack
Packaged 50/Rack, 300/CS
Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture
• Ideal for biological reaction applications and free gas exchange
• Aseptic conditions maintained through 0.22μm hydrophobic vented membrane
• Useful in the screening and optimization processes of suspension cells
• Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously
• Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization
Catalog No. 50-147-453 Supplier Quality Biological Inc Supplier No. P886229472
15 mL Bio-Reaction Tube with Vented Cap - Sterile, Foam Rack
Packaged 50/Rack, 300/CS
Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture
• Ideal for biological reaction applications and free gas exchange
• Aseptic conditions maintained through 0.22μm hydrophobic vented membrane
• Useful in the screening and optimization processes of suspension cells
• Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously
• Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization