Learn More
Bellco Glass, Inc. 15mL BioReaction Tube
SDP
Catalog No. 502330495
Encompass_Preferred
Bioreaction tubes offer a convenient and economical option for small scale high density suspension cell culture. Ideal for biological reaction applications and free gas exchange. Reaction parameters can be optimized using small amounts of materials maximizing the number of parallel experiments that can be performed simultaneously. Ideal for suspension cell culture bacterial growth seeding larger bioreactors supporting startup testing procedures for larger bioreactors and lyophilization.
Specifications
Aseptic conditions maintained through 0.22 hydrophobic vented membrane Useful in the screening and optimization processes of suspension cells Vent control labels for 50mL BioReaction tube caps cover 1 to 5 of the cap vent openings to control the amount of gas exchange Size 15mL Max Force 12000
Catalog No. 50-233-0495 Supplier Bellco Glass, Inc. Supplier No. 229471000
Bioreaction tubes offer a convenient and economical option for small scale high density suspension cell culture. Ideal for biological reaction applications and free gas exchange. Reaction parameters can be optimized using small amounts of materials maximizing the number of parallel experiments that can be performed simultaneously. Ideal for suspension cell culture bacterial growth seeding larger bioreactors supporting startup testing procedures for larger bioreactors and lyophilization.
Specifications
Aseptic conditions maintained through 0.22 hydrophobic vented membrane Useful in the screening and optimization processes of suspension cells Vent control labels for 50mL BioReaction tube caps cover 1 to 5 of the cap vent openings to control the amount of gas exchange Size 15mL Max Force 12000