Learn More
Diagenode Inc 15 ml Bioruptor Pico Tubes, 50pc
Catalog No. NC1670878
Encompass
The 15 ml Bioruptor Tubes strongly improve protein extraction efficiency. The tubes have been thoroughly validated for use on the Bioruptor Pico (B01060001) and are strongly recommended for extraction of proteins using this instrument. Recommended sample volume: 500 μl - 2 ml.
Catalog No. NC1670878 Supplier Diagenode Inc Supplier No. C30010017
The 15 ml Bioruptor Tubes strongly improve protein extraction efficiency. The tubes have been thoroughly validated for use on the Bioruptor Pico (B01060001) and are strongly recommended for extraction of proteins using this instrument. Recommended sample volume: 500 μl - 2 ml.