Learn More
Alkali Scientific 15mL Centrifuge Tubes, Conical, Foam Rack, Sterile, w/ Flat Blue Srew Cap, PP, 500/cs
SDP
Catalog No. NC1810213
Encompass_Preferred
Conical centrifuge tubes. High clarity polypropylene. Printed graduation marks. Sterile; Certified RNase, DNase & Pyrogen FREE.
Catalog No. NC1810213 Supplier Alkali Scientific Supplier No. C5600
Conical centrifuge tubes. High clarity polypropylene. Printed graduation marks. Sterile; Certified RNase, DNase & Pyrogen FREE.