Learn More
Quality Biological Inc 15 mL Centrifuge Tube & Cap - Non-sterile, (Caps and Tubes Packed Separately), Bag, 500 Tubes and 500 Caps/CS
SDP
Catalog No. 50147446
Encompass_Preferred
15 mL Centrifuge Tube & Cap - Non-sterile, (Caps and Tubes Packed Separately)
Packaged in Bags 500 Tubes and 500 Caps/CS
• Easy to read graduations and large white marking area.
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
• Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
Catalog No. 50-147-446 Supplier Quality Biological Inc Supplier No. P886229452
15 mL Centrifuge Tube & Cap - Non-sterile, (Caps and Tubes Packed Separately)
Packaged in Bags 500 Tubes and 500 Caps/CS
• Easy to read graduations and large white marking area.
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
• Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification