Learn More
CELLTREAT Scientific Products 15mL Centrifuge Tube - Plastic Rack, Sterile
SDP
Catalog No. 50607513
Encompass_Preferred
CELLTREAT Scientific Products 15mL Centrifuge Tube in Plastic Rack. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Plastic rack is manufactured from polypropylene and is autoclvable, reusable, and recyclable. Chemically resistant to drips. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily. RNase/DNase free and non-pyrogenic. Max force (RCF) g 12000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/plastic rack with 2 racks/bag, 300/case.
Catalog No. 50-607-513 Supplier CELLTREAT Scientific Products Supplier No. 229418
CELLTREAT Scientific Products 15mL Centrifuge Tube in Plastic Rack. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Plastic rack is manufactured from polypropylene and is autoclvable, reusable, and recyclable. Chemically resistant to drips. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily. RNase/DNase free and non-pyrogenic. Max force (RCF) g 12000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/plastic rack with 2 racks/bag, 300/case.