Learn More
Genesee Scientific Corporation 15 mL Conical Centrifuge Tubes, Leak-Proof Cap, Sterile (Gamma Irradiation), 500 Per Case
SDP
Catalog No. NC1118605
Encompass
Olympus 15 mL Centrifuge Tubes are designed for rigorous lab use, crafted from medical-grade virgin polypropylene to ensure chemical resistance and sample integrity. These sterile, leak-proof tubes come with a large white writing area for easy labeling and withstand forces up to 15,000 x g. Their autoclavable racks support eco-friendly practices, making them ideal for sensitive assays.
Catalog No. NC1118605 Supplier Genesee Scientific Corporation Supplier No. 28101
Olympus 15 mL Centrifuge Tubes are designed for rigorous lab use, crafted from medical-grade virgin polypropylene to ensure chemical resistance and sample integrity. These sterile, leak-proof tubes come with a large white writing area for easy labeling and withstand forces up to 15,000 x g. Their autoclavable racks support eco-friendly practices, making them ideal for sensitive assays.