Learn More
Chemglass Life Sciences 15ml Centrifuge Tube, PS Rack, Standard, Sterile, 50/Pk 500/Case
SDP
Catalog No. 501897793
Encompass_Preferred
Easy to see Black Graduation scaleLarge white writing areaEasy one-hand operationSterilized by E-Beam - DNase, RNase-free, non-pyrogenicRacked packaging is made using environmentally friendly re-usable racksHDPE material for the lid, PP material for the tubeRaw material complies with USP VI requirementsFlat markable cap with leak-proof sealMaximum centrifugation speeds: 12,000 x g
Catalog No. 50-189-7793 Supplier Chemglass Life Sciences Supplier No. CGN4301R15
Easy to see Black Graduation scaleLarge white writing areaEasy one-hand operationSterilized by E-Beam - DNase, RNase-free, non-pyrogenicRacked packaging is made using environmentally friendly re-usable racksHDPE material for the lid, PP material for the tubeRaw material complies with USP VI requirementsFlat markable cap with leak-proof sealMaximum centrifugation speeds: 12,000 x g