Learn More
Genesee Scientific Corporation 15 mL Conical Centrifuge Tubes, Sterile, Leak-Proof Cap
SDP
Catalog No. NC1890686
Encompass
These 15 mL conical centrifuge tubes from Genesee Scientific are designed for demanding lab applications, offering secure sample handling. Made from medical-grade polypropylene and sterilized by gamma irradiation, they are RNase, DNase, human DNA, and pyrogen-free. Featuring a leak-proof cap and high-contrast graduations, these tubes are ideal for precise volume measurement and can withstand centrifugal forces up to 15,000 x g, making them perfect for sensitive molecular biology work.
Catalog No. NC1890686 Supplier Genesee Scientific Corporation Supplier No. 28103
These 15 mL conical centrifuge tubes from Genesee Scientific are designed for demanding lab applications, offering secure sample handling. Made from medical-grade polypropylene and sterilized by gamma irradiation, they are RNase, DNase, human DNA, and pyrogen-free. Featuring a leak-proof cap and high-contrast graduations, these tubes are ideal for precise volume measurement and can withstand centrifugal forces up to 15,000 x g, making them perfect for sensitive molecular biology work.