Learn More
Phenomenex Inc 200mg PSA, 120mg C18, 0.6g MgSO4 (Anhydrous)
Catalog No. 502399271 Shop All Phenomenex Products
Encompass_Preferred
15mL Centrifugal Tube MS-9PA1219
Catalog No. 50-239-9271 Supplier Phenomenex Inc Supplier No. MS9PA1219
15mL Centrifugal Tube MS-9PA1219