Learn More
Phenomenex Inc 15mL Centrifugal Tube
Catalog No. 502397185 Shop All Phenomenex Products
Encompass_Preferred
15mL Centrifugal Tube 900mg PSA, 300mg C18, 0.9g MgSO4 (Anhydrous)
Catalog No. 50-239-7185 Supplier Phenomenex Inc Supplier No. MS9PA1215
15mL Centrifugal Tube 900mg PSA, 300mg C18, 0.9g MgSO4 (Anhydrous)