Learn More
CELLTREAT Scientific Products TUBE & CAP, 15mL Centrifuge Tube & Cap - Bags, Non-sterile (Caps and Tubes Packed Separately)
SDP
Catalog No. 50202018
Encompass_Preferred
CELLTREAT Scientific Products 15mL Tube & Cap Centrifuge Tube. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. RNase/DNase free and non-pyrogenic. Tube has max force (RCF) g of 12000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, tubes: 500/bag, caps: 500/bag, 500/case.
Catalog No. 50-202-018 Supplier CELLTREAT Scientific Products Supplier No. 229452
CELLTREAT Scientific Products 15mL Tube & Cap Centrifuge Tube. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. RNase/DNase free and non-pyrogenic. Tube has max force (RCF) g of 12000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, tubes: 500/bag, caps: 500/bag, 500/case.