Learn More
Bellco Glass, Inc. 15mL Centrifuge Tube
SDP
Catalog No. 502330070
Encompass_Preferred
DescriptionBlack tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foilLarge white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealConvenient zip-closure bag is a practical alternative to folding and taping opened bagsEasily refill reusable racks with tubes packed in bags Pack Description25/Re-sealable BagCase Quantity100SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorBlackPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume15mLGraduatedNoMax Force (RCF)g12000RNase/DNase FreeYes
Catalog No. 50-233-0070 Supplier Bellco Glass, Inc. Supplier No. 229431000
DescriptionBlack tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foilLarge white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealConvenient zip-closure bag is a practical alternative to folding and taping opened bagsEasily refill reusable racks with tubes packed in bags Pack Description25/Re-sealable BagCase Quantity100SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorBlackPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume15mLGraduatedNoMax Force (RCF)g12000RNase/DNase FreeYes