Learn More
Bellco Glass, Inc. 15mL Centrifuge Tube
SDP
Supplier:  Bellco Glass, Inc. 229412000
Encompass_Preferred
DescriptionGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsFoam rack is a sturdy alternative to disposable paperboard racks Pack Description50/Foam RackCase Quantity300SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: GreenPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume15mLGraduatedYesGraduation Increments (Subdivisions)0.5mLMax Force (RCF)g12000RNase/DNase FreeYes
Catalog No. 50-233-0592