Learn More
Quality Biological Inc 15 mL Centrifuge Tube only - Non-sterile, Bag - 500/CS
SDP
Catalog No. 50147444
Encompass_Preferred
15 mL Centrifuge Tube only - Non-sterile, Bag
Packaged 500/CS
• Polypropylene, 12,000 (RCF) g.
• Easy to read graduations and large white marking area.
Catalog No. 50-147-444 Supplier Quality Biological Inc Supplier No. P886229450
15 mL Centrifuge Tube only - Non-sterile, Bag
Packaged 500/CS
• Polypropylene, 12,000 (RCF) g.
• Easy to read graduations and large white marking area.