Learn More
CELLTREAT Scientific Products 15mL Centrifuge Tube - Black, Bag, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229431
Encompass_Preferred
CELLTREAT Scientific Products 15mL Black Centrifuge Tube. Black tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Easily refill reusable racks with tubes packed in bags. RNase/DNase free and non-pyrogenic. Max force (RCF) g 12000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable bag, 100/case.
Catalog No. 50-550-487