Learn More
Bellco Glass, Inc. 15mL Centrifuge Tube, Flip Top
SDP
Catalog No. 502330641
Encompass_Preferred
DescriptionGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsErgonomic flip top cap is designed for single handed operation Pack Description25/Re-sealable BagCase Quantity500SterileYesNon-PyrogenicYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: GreenPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume15mLGraduatedYesGraduation Increments (Subdivisions)0.5mLMax Force (RCF)g9400RNase/DNase FreeYes
Catalog No. 50-233-0641 Supplier Bellco Glass, Inc. Supplier No. 229417000
DescriptionGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsErgonomic flip top cap is designed for single handed operation Pack Description25/Re-sealable BagCase Quantity500SterileYesNon-PyrogenicYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: GreenPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume15mLGraduatedYesGraduation Increments (Subdivisions)0.5mLMax Force (RCF)g9400RNase/DNase FreeYes