Learn More
Chemglass Life Sciences Centrifuge Tube, 15mL, Graduated, Polypropylene, Sterile, Bulk Pack
SDP
Catalog No. 501897640
Encompass_Preferred
• Printed graduations (with marking area)• Polypropylene for the tube and polyethylene for the cap• Flat markable cap with leak-proof seal• One-hand operation design for the caps provides easy handling• Sterilized by E-Beam• Non-pyrogenic, DNase- and RNase-free• Packed in bags of 50• Max RCF (xg) 12,000
Catalog No. 50-189-7640 Supplier Chemglass Life Sciences Supplier No. CGN4303015
• Printed graduations (with marking area)• Polypropylene for the tube and polyethylene for the cap• Flat markable cap with leak-proof seal• One-hand operation design for the caps provides easy handling• Sterilized by E-Beam• Non-pyrogenic, DNase- and RNase-free• Packed in bags of 50• Max RCF (xg) 12,000