Learn More
CELLTREAT Scientific Products 15mL Centrifuge Tube, Metal Free - Paperboard Rack, Sterile, Case Quantity 500
SDP
Catalog No. 5000670039
Encompass_Preferred
Validated to contain less than 1ppb of more than 30 trace metal elements, ensuring samples are free of contaminants
Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
Large white marking area provides excellent contrast for identification
Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
Convenient zip-closure bag is a practical alternative to folding and taping opened bags
Easily refill reusable racks with tubes packed in bags
Catalog No. 50-006-70039 Supplier CELLTREAT Scientific Products Supplier No. 230481
Validated to contain less than 1ppb of more than 30 trace metal elements, ensuring samples are free of contaminants
Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
Large white marking area provides excellent contrast for identification
Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
Convenient zip-closure bag is a practical alternative to folding and taping opened bags
Easily refill reusable racks with tubes packed in bags