Learn More
Bellco Glass, Inc. 15mL Centrifuge Tube, Snap
SDP
Catalog No. 502330274
Encompass_Preferred
DescriptionSingle piece lid and tube available in conical and self-standing bottomsTab on lid enables convenient one-handed openingLid audibly snaps closed, and opens with a ‘pop!’ so you know when it is sealedIdeal for sample storage, collection, and experimental procedures Pack Description50/Re-sealable BagCase Quantity500SterileYesNon-PyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume15mLGraduatedYesGraduation Increments (Subdivisions)0.5mLMax Force (RCF)g5500RNase/iDNase FreeYesle Bag
Catalog No. 50-233-0274 Supplier Bellco Glass, Inc. Supplier No. 229496000
DescriptionSingle piece lid and tube available in conical and self-standing bottomsTab on lid enables convenient one-handed openingLid audibly snaps closed, and opens with a ‘pop!’ so you know when it is sealedIdeal for sample storage, collection, and experimental procedures Pack Description50/Re-sealable BagCase Quantity500SterileYesNon-PyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume15mLGraduatedYesGraduation Increments (Subdivisions)0.5mLMax Force (RCF)g5500RNase/iDNase FreeYesle Bag