Learn More
Azzota Corp 15Ml Centrifuge Tubes, Made Of High-Clarity Polypropylene. Allowable Temperature From -80Deg.C To 120Deg.C. Can Sustain 12,000G, Conical Bottom, 50 Tubes And Caps Per Bag.
Catalog No. 507532951
Encompass_Preferred
15ml Centrifuge Tubes, Made of high-clarity polypropylene. Allowable temperature from -80deg.C to 120deg.C. Can sustain 12,000g, Conical Bottom, 50 tubes and caps per bag.
Catalog No. 50-753-2951 Supplier Azzota Corp Supplier No. CT15
15ml Centrifuge Tubes, Made of high-clarity polypropylene. Allowable temperature from -80deg.C to 120deg.C. Can sustain 12,000g, Conical Bottom, 50 tubes and caps per bag.