Learn More
CELLTREAT Scientific Products 15mL Centrifuge Tube, Snap-Pop Lid, Self-Standing - Bag, Sterile, 500/CS
SDP
Catalog No. 501535100
Encompass_Preferred
CELLTREAT Scientific Products 15mL Snap-Pop Lid Centrifuge Tube. Single piece lid and tube with self-standing bottom. Tab on lid enables convenient one-handed opening. Lid audibly snaps closed, and opens with a 'pop!' so you know when it is sealed. Ideal for sample storage, collection, and experimental procedures. Tube features molded graduation marks at 0.5mL increments, numbered every 1mL. 15mL maximum volume. Temperature range of -80¿C to 120¿C. Max RCF: 1,000g. Tube and lid manufactured from virgin polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 50/re-sealable bag, 500/case.
Catalog No. 50-153-5100 Supplier CELLTREAT Scientific Products Supplier No. 229497
CELLTREAT Scientific Products 15mL Snap-Pop Lid Centrifuge Tube. Single piece lid and tube with self-standing bottom. Tab on lid enables convenient one-handed opening. Lid audibly snaps closed, and opens with a 'pop!' so you know when it is sealed. Ideal for sample storage, collection, and experimental procedures. Tube features molded graduation marks at 0.5mL increments, numbered every 1mL. 15mL maximum volume. Temperature range of -80¿C to 120¿C. Max RCF: 1,000g. Tube and lid manufactured from virgin polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 50/re-sealable bag, 500/case.