Learn More
Alkali Scientific ASI™ 15mL Dialysis Chamber, PC/RC, 13-15mL, 10/Cs, 14k, White
SDP
Catalog No. 503026629
Encompass_Preferred
ASI™ Dialysis Chamber is used for various dialysis applications such as desalting, buffer exchange, labeling, reagent removal, drug binding experiment, cell growth and supply, virus purifications, blood transformation, etc. It is configured to rapidly and effectively dialysis a wide range of 15mL volume and guarantees 95 ~ 98% higher recovery rate than Dialysis tubing type. ▪ Excellent pH and chemical stability (pH 5 ~ 9)▪ Membrane composition: Regenerated Cellulose (RC)▪ Membrane Weight Cut-Off (MWCO): 3.5K, 7K 14K
Catalog No. 50-302-6629 Supplier Alkali Scientific Supplier No. DC015W
ASI™ Dialysis Chamber is used for various dialysis applications such as desalting, buffer exchange, labeling, reagent removal, drug binding experiment, cell growth and supply, virus purifications, blood transformation, etc. It is configured to rapidly and effectively dialysis a wide range of 15mL volume and guarantees 95 ~ 98% higher recovery rate than Dialysis tubing type. ▪ Excellent pH and chemical stability (pH 5 ~ 9)▪ Membrane composition: Regenerated Cellulose (RC)▪ Membrane Weight Cut-Off (MWCO): 3.5K, 7K 14K