Learn More
Chemglass Life Sciences 15mL Filter Funnel, Buchner, 19/22 Lower Vacuum Assembly, Coarse Frit
SDP
Supplier:  Chemglass Life Sciences CG140645
Encompass_Preferred
15mL Filter Funnel, Buchner, 19/22 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140645