Learn More
Ace Glass, Inc. 15mL volumetric flask features a 12/18 joint
SDP
Catalog No. 509557602
Encompass_Preferred
15mL volumetric flask features a 12/18 joint with an approximate 10mm opening. Cylindrical body allows for better mixing and drainage as well as ease of withdrawal using pipets. Its wide base offers stability, and the upper surface of the base is ground to allow pencil markings. Flask is for Class A tolerances and is supplied with one stopper.
Catalog No. 50-955-7602 Supplier Ace Glass, Inc. Supplier No. 712415
15mL volumetric flask features a 12/18 joint with an approximate 10mm opening. Cylindrical body allows for better mixing and drainage as well as ease of withdrawal using pipets. Its wide base offers stability, and the upper surface of the base is ground to allow pencil markings. Flask is for Class A tolerances and is supplied with one stopper.