Learn More
Foxx Life Sciences 15mL Narrow Mouth HDPE Reagent Bottles, 72CS
GREENER_CHOICE
Catalog No. 5000846656
Encompass_Preferred
EZLabpure™  15mL HDPE Reagent Bottles Non Sterile provide safe, leak-proof containment for lab chemicals, solution & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these lightweight, shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth HDPE Reagent Bottle ensures precision dispensing, controlled pouring & low spillage. HDPE chemical resistant reagent bottles offer excellent chemical resistance against acids, bases & alcohols. Reagent Bottles are freezer-safe for protecting sensitive samples during cold storage.
Features
High impact strength & chemical resistance
Freezer safe reagent bottles are non-autoclavable & rated for temperatures down to -100°C
HDPE storage bottles are ideal for mild acids, bases, and alcohols
Lightweight and durable, HDPE storage reagent bottles are designed for long-term storage
Catalog No. 50-008-46656 Supplier Foxx Life Sciences Supplier No. 7621511FLS
EZLabpure™  15mL HDPE Reagent Bottles Non Sterile provide safe, leak-proof containment for lab chemicals, solution & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these lightweight, shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth HDPE Reagent Bottle ensures precision dispensing, controlled pouring & low spillage. HDPE chemical resistant reagent bottles offer excellent chemical resistance against acids, bases & alcohols. Reagent Bottles are freezer-safe for protecting sensitive samples during cold storage.
Features
High impact strength & chemical resistance
Freezer safe reagent bottles are non-autoclavable & rated for temperatures down to -100°C
HDPE storage bottles are ideal for mild acids, bases, and alcohols
Lightweight and durable, HDPE storage reagent bottles are designed for long-term storage