Learn More
Bellco Glass, Inc. 1.5mL Micro Centrifuge Tube
SDP
Catalog No. 502330489
Encompass_Preferred
DescriptionTubes have frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handBlack tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil Pack Description500/Re-sealable BagCase Quantity1000SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorBlackPrimary MaterialPolypropyleneMaximum Volume1.5mLGraduatedYesGraduation Increments (Subdivisions)0.25mLMax Force (RCF)g25000RNase/DNase FreeYes
Catalog No. 50-233-0489 Supplier Bellco Glass, Inc. Supplier No. 229437000
DescriptionTubes have frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handBlack tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil Pack Description500/Re-sealable BagCase Quantity1000SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorBlackPrimary MaterialPolypropyleneMaximum Volume1.5mLGraduatedYesGraduation Increments (Subdivisions)0.25mLMax Force (RCF)g25000RNase/DNase FreeYes