Learn More
Ace Glass, Inc. 15mL glass Pestle with 10mm rod,
SDP
Catalog No. 50974568
Encompass_Preferred
15mL glass Pestle with 10mm rod, for use with 15mL tapered tube (50979772) as a tissue grinder. The rod can be used with a 509558293 Flex-Grip Chuck.
This item is not returnable. View return policy
Catalog No. 50-974-568 Supplier Ace Glass, Inc. Supplier No. 832518
15mL glass Pestle with 10mm rod, for use with 15mL tapered tube (50979772) as a tissue grinder. The rod can be used with a 509558293 Flex-Grip Chuck.